Anti-C11orf24 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP49708_P050
AVIVARP49708_P050EA 401 EUR
AVIVARP49708_P050
Anti-C11orf24 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: C11orf24 (chromosome 11 open reading frame 24)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human