Anti-CYP3A43 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP49757_P050
AVIVARP49757_P050EA 401 EUR
AVIVARP49757_P050
Anti-CYP3A43 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP3A43 (cytochrome P450, family 3, subfamily A, polypeptide 43)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human