Anti-TMEM132D Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP50299_P050
AVIVARP50299_P050EA 401 EUR
AVIVARP50299_P050
Anti-TMEM132D Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMEM132D (transmembrane protein 132D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen