Anti-EMID1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP50301_P050
AVIVARP50301_P050EA 401 EUR
AVIVARP50301_P050
Anti-EMID1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EMID1 (EMI domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen