Anti-TRAPPC2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP50567_P050
AVIVARP50567_P050EA 401 EUR
AVIVARP50567_P050
Anti-TRAPPC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRAPPC2 (trafficking protein particle complex 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat