Anti-ZNF418 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP50765_P050
AVIVARP50765_P050EA 401 EUR
AVIVARP50765_P050
Anti-ZNF418 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF418 (zinc finger protein 418)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR