Anti-TEC Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP51209_P050
AVIVARP51209_P050EA 401 EUR
AVIVARP51209_P050
Anti-TEC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TEC (tec protein tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen