Anti-PEG10 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP51313_P050
AVIVARP51313_P050EA 401 EUR
AVIVARP51313_P050
Anti-PEG10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PEG10 (paternally expressed 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen