Anti-PPIA Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP51353_P050
AVIVARP51353_P050EA 401 EUR
AVIVARP51353_P050
Anti-PPIA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPIA (peptidylprolyl isomerase A (cyclophilin A))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen