Anti-OAS1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP51360_P050
AVIVARP51360_P050EA 401 EUR
AVIVARP51360_P050
Anti-OAS1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OAS1 (2'-5'-oligoadenylate synthetase 1, 40/46kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human