Anti-CTF1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP51944_P050
AVIVARP51944_P050EA 401 EUR
AVIVARP51944_P050
Anti-CTF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CTF1 (cardiotrophin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen