Anti-CHN1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP51973_P050
AVIVARP51973_P050EA 401 EUR
AVIVARP51973_P050
Anti-CHN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHN1 (chimerin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen