Anti-CHN1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP51974_P050
AVIVARP51974_P050EA 401 EUR
AVIVARP51974_P050
Anti-CHN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHN1 (chimerin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen