Anti-AP4S1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP52284_P050
AVIVARP52284_P050EA 401 EUR
AVIVARP52284_P050
Anti-AP4S1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AP4S1 (adaptor-related protein complex 4, sigma 1 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR