Anti-PLEKHH2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP53218_P050
AVIVARP53218_P050EA 401 EUR
AVIVARP53218_P050
Anti-PLEKHH2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLEKHH2 (pleckstrin homology domain containing, family H (with MyTH4 domain) member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR