Anti-MISP Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP53237_P050
AVIVARP53237_P050EA 401 EUR
AVIVARP53237_P050
Anti-MISP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MISP (mitotic spindle positioning)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Guinea pig