Anti-MCMDC2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP53263_P050
AVIVARP53263_P050EA 401 EUR
AVIVARP53263_P050
Anti-MCMDC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MCMDC2 (minichromosome maintenance domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Guinea pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR