Anti-GSTM3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP53562_P050
AVIVARP53562_P050EA 401 EUR
AVIVARP53562_P050
Anti-GSTM3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GSTM3 (glutathione S-transferase mu 3 (brain))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen