Anti-GRK4 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP53566_P050
AVIVARP53566_P050EA 401 EUR
AVIVARP53566_P050
Anti-GRK4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GRK4 (G protein-coupled receptor kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen