Anti-CHKA Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP53583_P050
AVIVARP53583_P050EA 401 EUR
AVIVARP53583_P050
Anti-CHKA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHKA (choline kinase alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen