Anti-PCYT2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP53606_P050
AVIVARP53606_P050EA 401 EUR
AVIVARP53606_P050
Anti-PCYT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PCYT2 (phosphate cytidylyltransferase 2, ethanolamine)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen