Anti-TTC12 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP53725_P050
AVIVARP53725_P050EA 401 EUR
AVIVARP53725_P050
Anti-TTC12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TTC12 (tetratricopeptide repeat domain 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR