Anti-GTF2A1L Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP53836_P050
AVIVARP53836_P050EA 401 EUR
AVIVARP53836_P050
Anti-GTF2A1L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GTF2A1L (general transcription factor IIA, 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen