Anti-ATG10 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP54274_P050
AVIVARP54274_P050EA 401 EUR
AVIVARP54274_P050
Anti-ATG10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATG10 (autophagy related 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen