Anti-D9Ertd402e Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP54464_P050
AVIVARP54464_P050EA 401 EUR
AVIVARP54464_P050
Anti-D9Ertd402e Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: D9Ertd402e
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse