Anti-NARF Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP54490_P050
AVIVARP54490_P050EA 401 EUR
AVIVARP54490_P050
Anti-NARF Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NARF (nuclear prelamin A recognition factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen