Anti-SFN Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP54791_P050
AVIVARP54791_P050EA 401 EUR
AVIVARP54791_P050
Anti-SFN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SFN (stratifin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen