Anti-BLOC1S6 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP54890_P050
AVIVARP54890_P050EA 401 EUR
AVIVARP54890_P050
Anti-BLOC1S6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BLOC1S6 (biogenesis of lysosomal organelles complex-1, subunit 6, pallidin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR