Anti-WBP2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP54911_P050
AVIVARP54911_P050EA 401 EUR
AVIVARP54911_P050
Anti-WBP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: WBP2 (WW domain binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR