Anti-PIK3R4 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP55087_P050
AVIVARP55087_P050EA 401 EUR
AVIVARP55087_P050
Anti-PIK3R4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIK3R4 (phosphoinositide-3-kinase, regulatory subunit 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR