Anti-FBXO28 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP55192_P050
AVIVARP55192_P050EA 401 EUR
AVIVARP55192_P050
Anti-FBXO28 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FBXO28 (F-box protein 28)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen