Anti-SAMHD1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP55262_P050
AVIVARP55262_P050EA 401 EUR
AVIVARP55262_P050
Anti-SAMHD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SAMHD1 (SAM domain and HD domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen