Anti-IRF2BP1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP55314_P050
AVIVARP55314_P050EA 401 EUR
AVIVARP55314_P050
Anti-IRF2BP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IRF2BP1 (interferon regulatory factor 2 binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen