Anti-LYSMD2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP55592_P050
AVIVARP55592_P050EA 401 EUR
AVIVARP55592_P050
Anti-LYSMD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LYSMD2 (LysM, putative peptidoglycan-binding, domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen