Anti-CCDC96 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP55593_P050
AVIVARP55593_P050EA 401 EUR
AVIVARP55593_P050
Anti-CCDC96 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCDC96 (coiled-coil domain containing 96)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR