Anti-PMS2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP56117_P050
AVIVARP56117_P050EA 401 EUR
AVIVARP56117_P050
Anti-PMS2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PMS2 (PMS2 postmeiotic segregation increased 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen