Anti-SH3GL3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP56551_P050
AVIVARP56551_P050EA 401 EUR
AVIVARP56551_P050
Anti-SH3GL3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SH3GL3 (SH3-domain GRB2-like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen