Anti-PRKCH Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP56704_P050
AVIVARP56704_P050EA 401 EUR
AVIVARP56704_P050
Anti-PRKCH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRKCH (protein kinase C, eta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen