Anti-PSMC4 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP56716_P050
AVIVARP56716_P050EA 401 EUR
AVIVARP56716_P050
Anti-PSMC4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PSMC4 (proteasome (prosome, macropain) 26S subunit, ATPase, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen