Anti-RLN1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP56721_P050
AVIVARP56721_P050EA 401 EUR
AVIVARP56721_P050
Anti-RLN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RLN1 (relaxin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human