Anti-MED31 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP56821_P050
AVIVARP56821_P050EA 401 EUR
AVIVARP56821_P050
Anti-MED31 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MED31 (mediator complex subunit 31)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat