Anti-OGDHL Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP57175_P050
AVIVARP57175_P050EA 401 EUR
AVIVARP57175_P050
Anti-OGDHL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OGDHL (oxoglutarate dehydrogenase-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen