Anti-TUBA8 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP57298_P050
AVIVARP57298_P050EA 401 EUR
AVIVARP57298_P050
Anti-TUBA8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TUBA8 (tubulin, alpha 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR