Anti-EXOC6 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP57319_P050
AVIVARP57319_P050EA 401 EUR
AVIVARP57319_P050
Anti-EXOC6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EXOC6 (exocyst complex component 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen