Anti-ACBD3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP57649_P050
AVIVARP57649_P050EA 401 EUR
AVIVARP57649_P050
Anti-ACBD3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACBD3 (acyl-CoA binding domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen