Anti-PDE8A Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP57753_P050
AVIVARP57753_P050EA 401 EUR
AVIVARP57753_P050
Anti-PDE8A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PDE8A (phosphodiesterase 8A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen