Anti-GRM6 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP58004_P050
AVIVARP58004_P050EA 401 EUR
AVIVARP58004_P050
Anti-GRM6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GRM6 (glutamate receptor, metabotropic 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen