Anti-ACD Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP58253_P050
AVIVARP58253_P050EA 401 EUR
AVIVARP58253_P050
Anti-ACD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACD (adrenocortical dysplasia homolog (mouse))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen