Anti-H2AFX Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP58281_P050
AVIVARP58281_P050EA 401 EUR
AVIVARP58281_P050
Anti-H2AFX Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H2AFX (H2A histone family, member X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen