Anti-PTGES3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP58319_P050
AVIVARP58319_P050EA 401 EUR
AVIVARP58319_P050
Anti-PTGES3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTGES3 (prostaglandin E synthase 3 (cytosolic))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen